Gene Transcripts

Gene Symbol
PLXDC2
Entrez Gene ID
84898
Gene Name
plexin domain containing 2
Gene Aliases
TEM7R
Chromosome Location
Chr. 10: 19816215 - 20289856 on Build GRCh38
UniGene ID
Hs.658134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282736.1 NP_001269665.1 10, 11 1763
NM_032812.8 NP_116201.7 11, 12 1910
XM_011519750.2 XP_011518052.1 11, 12 2138
GenBank mRNA AF378757.1 - 11, 12 1282
AK027529.1 - 5, 6 562
AK304591.1 - 12, 13 1794
AY358486.1 - 11, 12 1910
BC012885.1 - 10, 11 1369 3865073

View Full Product Details