Gene Transcripts

Gene Symbol
ZCCHC3
Entrez Gene ID
85364
Gene Name
zinc finger CCHC-type containing 3
Gene Aliases
C20orf99
Chromosome Location
Chr. 20: 297563 - 300321 on Build GRCh38
UniGene ID
Hs.28608
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033089.6 NP_149080.2 1 1405
GenBank mRNA AK025768.1 - 2 690
BC069238.1 - 1 1405 6287290
BC107584.1 - 2 907 6457775

View Full Product Details