Gene Transcripts

Gene Symbol
FHDC1
Entrez Gene ID
85462
Gene Name
FH2 domain containing 1
Gene Aliases
Chromosome Location
Chr. 4: 152911570 - 152979696 on Build GRCh38
UniGene ID
Hs.132629
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033393.2 NP_203751.2 2 607
XM_005263319.3 XP_005263376.1 3 748
XM_006714393.3 XP_006714456.1 3 757
XM_011532388.1 XP_011530690.1 5 1239
XM_011532389.1 XP_011530691.1 3 1353
GenBank mRNA AB051514.1 - 2 494

View Full Product Details