Gene Transcripts
- Gene Symbol
- PAGE5
- Entrez Gene ID
- 90737
- Gene Name
- PAGE family member 5
- Gene Aliases
- CT16, CT16.1, CT16.2, GAGEE1, PAGE-5
- Chromosome Location
- Chr. X: 55220347 - 55224108 on Build GRCh38
- UniGene ID
- Hs.668491
- Species
- Homo sapiens
- Gene Symbol
- LOC100421746
- Entrez Gene ID
- 100421746
- Gene Name
- PAGE family member 5 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. X: 55280088 - 55288786 on Build GRCh38
- UniGene ID
- Hs.662612
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_130179.1 |
|
5 |
482 |
|
View Full Product Details