Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318785.1 | NP_001305714.1 | 10 | 1132 | |
NM_020320.4 | NP_064716.2 | 11 | 967 | ||
NR_134857.1 | 11 | 967 | |||
XM_011535948.2 | XP_011534250.1 | 11 | 952 | ||
XM_011535949.2 | XP_011534251.1 | 11 | 952 | ||
XM_017011072.1 | XP_016866561.1 | 11 | 1133 | ||
XM_017011073.1 | XP_016866562.1 | 12 | 1351 | ||
XM_017011074.1 | XP_016866563.1 | 11 | 954 | ||
XM_017011075.1 | XP_016866564.1 | 10 | 1077 | ||
XM_017011076.1 | XP_016866565.1 | 12 | 1295 | ||
XM_017011077.1 | XP_016866566.1 | 10 | 854 | ||
XM_017011078.1 | XP_016866567.1 | 10 | 898 | ||
XM_017011079.1 | XP_016866568.1 | 11 | 1133 | ||
XM_017011080.1 | XP_016866569.1 | 12 | 1295 | ||
XM_017011081.1 | XP_016866570.1 | 10 | 898 | ||
GenBank mRNA | AK023550.1 | - | 11 | 910 | |
AK093934.1 | - | 11 | 1340 | ||
AK315669.1 | - | 11 | 904 | ||
BC010420.1 | - | 11 | 938 | 3613406 | |
BC105784.1 | - | 10 | 1071 |