Gene Transcripts

Gene Symbol
IDI2
Entrez Gene ID
91734
Gene Name
isopentenyl-diphosphate delta isomerase 2
Gene Aliases
IPPI2
Chromosome Location
Chr. 10: 1018907 - 1025859 on Build GRCh38
UniGene ID
Hs.591325
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033261.2 NP_150286.1 5 562
GenBank mRNA AF271725.1 - 4 538
AK056950.1 - 5 562
BC017778.1 - 5 561 4291149

View Full Product Details