Gene Transcripts

Gene Symbol
AZIN2
Entrez Gene ID
113451
Gene Name
antizyme inhibitor 2
Gene Aliases
ADC, AZI2, AZIB1, ODC-p, ODC1L, ODCp
Chromosome Location
Chr. 1: 33081110 - 33162288 on Build GRCh38
UniGene ID
Hs.101807
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293562.1 NP_001280491.1 9 1363
NM_001301823.1 NP_001288752.1 7 1266
NM_001301824.1 NP_001288753.1 6 1092
NM_001301825.1 NP_001288754.1 7 1123
NM_001301826.1 NP_001288755.1 7 1063
NM_052998.3 NP_443724.1 10 1578
NR_126031.1 7 1100
XM_005270404.2 XP_005270461.1 9 1325
XM_005270406.1 XP_005270463.1 8 1192
XM_005270407.1 XP_005270464.1 7 962
XM_006710312.1 XP_006710375.1 7 1010
XM_011540557.1 XP_011538859.1 8 1144
XM_011540563.1 XP_011538865.1 9 1323
XM_011540566.1 XP_011538868.1 9 1334
XM_017000167.1 XP_016855656.1 9 1323
XM_017000168.1 XP_016855657.1 10 1448
XM_017000169.1 XP_016855658.1 10 1356
XM_017000170.1 XP_016855659.1 9 1350
XM_017000171.1 XP_016855660.1 10 1436
XM_017000172.1 XP_016855661.1 10 1353
XM_017000173.1 XP_016855662.1 8 1029
XM_017000174.1 XP_016855663.1 9 1334
XM_017000175.1 XP_016855664.1 8 1148
XM_017000176.1 XP_016855665.1 8 1122
XM_017000177.1 XP_016855666.1 8 1180
XM_017000178.1 XP_016855667.1 8 1133
GenBank mRNA AB075825.1 - 6 3181
AK057051.1 - 6 1066
AK095127.1 - 7 1247
AK122697.1 - 6 1971
AK315677.1 - 9 1325
AL832640.2 - 9 2114
AY050634.1 - 7 991
AY050635.1 - 7 1051
AY050636.1 - 7 1027
AY050637.1 - 7 991
AY050638.1 - 4 517
AY325129.1 - 9 1309
BC010449.1 - 9 1309 4156927
BC028128.1 - 10 1578 5164244

View Full Product Details