Gene Transcripts

Gene Symbol
C8orf34
Entrez Gene ID
116328
Gene Name
chromosome 8 open reading frame 34
Gene Aliases
VEST-1, VEST1
Chromosome Location
Chr. 8: 68330722 - 68819023 on Build GRCh38
UniGene ID
Hs.491941
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195639.1 NP_001182568.1 6 1197
NM_052958.2 NP_443190.2 6 1197
XM_011517445.2 XP_011515747.1 6 1169
XM_011517446.2 XP_011515748.1 6 1169
XM_011517448.2 XP_011515750.1 5 1040
XM_011517449.2 XP_011515751.1 6 1169
XM_011517450.2 XP_011515752.1 6 1169
XM_017013019.1 XP_016868508.1 5 1040
GenBank mRNA AB056652.1 - 5 2165
AK291436.1 - 6 669
AL834454.1 - 6 694
BC041961.1 - 6 1197 5302156

View Full Product Details