Gene Transcripts
- Gene Symbol
- TRIM6
- Entrez Gene ID
- 117854
- Gene Name
- tripartite motif containing 6
- Gene Aliases
- RNF89
- Chromosome Location
- Chr. 11: 5596101 - 5612958 on Build GRCh38
- UniGene ID
- Hs.729048
- Species
- Homo sapiens
- Gene Symbol
- TRIM6-TRIM34
- Entrez Gene ID
- 445372
- Gene Name
- TRIM6-TRIM34 readthrough
- Gene Aliases
- IFP1, RNF21, TRIM34
- Chromosome Location
- Chr. 11: 5596635 - 5644398 on Build GRCh38
- UniGene ID
- Hs.729048
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CX760618.1 |
- |
5 |
724 |
|
View Full Product Details