Gene Transcripts

Gene Symbol
B4GALNT2
Entrez Gene ID
124872
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 2
Gene Aliases
B4GALT, GALGT2
Chromosome Location
Chr. 17: 49120347 - 49170722 on Build GRCh38
UniGene ID
Hs.374679
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF510036.1 - 8 1169
AJ517770.1 - 8 1169
AK302876.1 - 8 967
BC113675.1 - 8 1163 8322727
BC113677.1 - 8 1163 8322729
S83275.1 - 2 231

View Full Product Details