Gene Transcripts

Gene Symbol
FAM213B
Entrez Gene ID
127281
Gene Name
family with sequence similarity 213 member B
Gene Aliases
C1orf93
Chromosome Location
Chr. 1: 2586460 - 2591469 on Build GRCh38
UniGene ID
Hs.462033
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195736.1 NP_001182665.1 7 1723
NM_001195737.1 NP_001182666.1 7 1657
NM_001195738.1 NP_001182667.1 7 1637
NM_001195740.1 NP_001182669.1 6 1561
NM_001195741.1 NP_001182670.1 6 1558
NM_152371.3 NP_689584.2 7 1669
NR_036637.1 7 1602
NR_036638.1 7 1570
XM_006710354.3 XP_006710417.1 7 1657
XM_011540664.1 XP_011538966.1 7 1713
XM_011540665.1 XP_011538967.1 7 1681
XM_011540666.1 XP_011538968.1 7 1661
GenBank mRNA AF425266.1 - 7 1614
AK057027.1 - 7 1654
AK075273.1 - 7 1589
AK094901.1 - 6 1471
BC022547.1 - 7 1609 4812314
CR749706.1 - 2 1401

View Full Product Details