Gene Transcripts

Gene Symbol
NUP35
Entrez Gene ID
129401
Gene Name
nucleoporin 35
Gene Aliases
MP-44, MP44, NP44, NUP53
Chromosome Location
Chr. 2: 183117490 - 183161684 on Build GRCh38
UniGene ID
Hs.180591
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287584.1 NP_001274513.1 8 847
NM_001287585.1 NP_001274514.1 8 867
NM_138285.4 NP_612142.2 7 744
NR_109856.1 6 602
XM_006712254.2 XP_006712317.1 7 709
XM_011510576.2 XP_011508878.1 9 975
XM_011510577.2 XP_011508879.1 9 1162
XM_017003308.1 XP_016858797.1 7 900
GenBank mRNA AF411516.1 - 7 744
AF514993.1 - 7 701
AK289601.1 - 6 545
AK290760.1 - 7 656
AK298199.1 - 8 777
AK302350.1 - 8 816
BC035597.1 - 7 668
BC047029.1 - 7 731 5541023
BC061698.1 - 7 698 4132440
BI818393.1 - 7 707
BI821231.1 - 6 538

View Full Product Details