Gene Transcripts

Gene Symbol
NCOA7
Entrez Gene ID
135112
Gene Name
nuclear receptor coactivator 7
Gene Aliases
ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3
Chromosome Location
Chr. 6: 125780948 - 125932030 on Build GRCh38
UniGene ID
Hs.171426
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122842.2 NP_001116314.1 7 987
NM_001199619.1 NP_001186548.1 8 970
NM_001199620.1 NP_001186549.1 9 1088
NM_001199621.1 NP_001186550.1 6 766
NM_181782.4 NP_861447.3 7 987
XM_005266822.4 XP_005266879.1 7 1017
XM_006715340.3 XP_006715403.1 7 944
XM_011535455.2 XP_011533757.1 7 892
XM_017010269.1 XP_016865758.1 7 703
XM_017010270.1 XP_016865759.1 8 949
XM_017010271.1 XP_016865760.1 7 947
XM_017010272.1 XP_016865761.1 7 800
XM_017010273.1 XP_016865762.1 7 1017
XM_017010274.1 XP_016865763.1 7 1106
GenBank mRNA AB074156.1 - 6 515
AB074157.1 - 7 787
AF493978.1 - 8 970
AK127512.1 - 7 797
AK294558.1 - 6 564
AK307395.1 - 8 949
AL832621.2 - 7 780
AL832628.2 - 7 744
AL834442.1 - 5 408
BC036461.1 - 7 815
BC137094.1 - 9 1004 9021091
BC137095.1 - 8 886 9021092
BX537385.1 - 7 782

View Full Product Details