Gene Transcripts

Gene Symbol
BRI3BP
Entrez Gene ID
140707
Gene Name
BRI3 binding protein
Gene Aliases
BNAS1, HCCR-1, HCCR-2, HCCRBP-1, KG19
Chromosome Location
Chr. 12: 124993648 - 125051845 on Build GRCh38
UniGene ID
Hs.596464
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080626.5 NP_542193.3 3 988
GenBank mRNA AF283671.2 - 3 727
AF284094.1 - 3 932
AK055703.1 - 1 509
BC034525.1 - 3 965 4778946

View Full Product Details