Gene Transcripts

Gene Symbol
OR52I2
Entrez Gene ID
143502
Gene Name
olfactory receptor family 52 subfamily I member 2
Gene Aliases
OR11-12, OR52I1
Chromosome Location
Chr. 11: 4586791 - 4587905 on Build GRCh38
UniGene ID
Hs.553607
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005170.2 NP_001005170.1 1 1034
GenBank mRNA BC136921.1 - 1 1034
BC151144.1 - 1 1034 9020913

View Full Product Details