Gene Transcripts

Gene Symbol
C16orf89
Entrez Gene ID
146556
Gene Name
chromosome 16 open reading frame 89
Gene Aliases
Chromosome Location
Chr. 16: 5042772 - 5066145 on Build GRCh38
UniGene ID
Hs.11782
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098514.2 NP_001091984.2 2, 3 589
NM_152459.4 NP_689672.4 2, 3 589
XM_005255143.3 XP_005255200.2 2, 3 415
XM_017022972.1 XP_016878461.1 2, 3 415
XM_017022973.1 XP_016878462.1 2, 3 415
XM_017022974.1 XP_016878463.1 2, 3 415
GenBank mRNA AK075242.1 - 2, 3 415
AK300708.1 - 2, 3 363
AY358483.1 - 2, 3 554
BC033681.1 - 2, 3 484 5190553

View Full Product Details