Gene Transcripts

Gene Symbol
CCDC27
Entrez Gene ID
148870
Gene Name
coiled-coil domain containing 27
Gene Aliases
Chromosome Location
Chr. 1: 3752398 - 3771645 on Build GRCh38
UniGene ID
Hs.348700
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152492.2 NP_689705.2 10, 11 1831
XM_011540780.1 XP_011539082.1 10, 11 1834
XM_017000402.1 XP_016855891.1 9, 10 2065
XM_017000403.1 XP_016855892.1 9, 10 2065
XM_017000404.1 XP_016855893.1 9, 10 2065
XM_017000406.1 XP_016855895.1 10, 11 1879
GenBank mRNA AK057387.1 - 10, 11 1834
BC112108.1 - 10, 11 1832 8327576
BC112112.1 - 10, 11 1831 8327580

View Full Product Details