Gene Transcripts
- Gene Symbol
- BTLA
- Entrez Gene ID
- 151888
- Gene Name
- B and T lymphocyte associated
- Gene Aliases
- BTLA1, CD272
- Chromosome Location
- Chr. 3: 112458790 - 112499756 on Build GRCh38
- UniGene ID
- Hs.445162
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001085357.1 |
NP_001078826.1
|
3, 4 |
645 |
|
NM_181780.3 |
NP_861445.3
|
4, 5 |
789 |
|
XM_011512447.2 |
XP_011510749.1
|
5, 6 |
901 |
|
XM_017005748.1 |
XP_016861237.1
|
5, 6 |
864 |
|
GenBank mRNA |
AJ717664.1 |
- |
4, 5 |
586 |
|
AK131204.1 |
- |
4, 5 |
578 |
|
AK292692.1 |
- |
4, 5 |
768 |
|
AY599411.1 |
- |
3, 4 |
442 |
|
AY606805.1 |
- |
4, 5 |
586 |
|
BC107091.1 |
- |
3, 4 |
442 |
40015588 |
BC107092.1 |
- |
4, 5 |
584 |
|
DQ198368.1 |
- |
3, 4 |
442 |
|
View Full Product Details