Gene Transcripts

Gene Symbol
CCDC12
Entrez Gene ID
151903
Gene Name
coiled-coil domain containing 12
Gene Aliases
Chromosome Location
Chr. 3: 46917905 - 46982031 on Build GRCh38
UniGene ID
Hs.631918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277074.1 NP_001264003.1 2 162
NM_144716.5 NP_653317.2 3 317
NR_102269.1 2 180
XM_011533390.2 XP_011531692.1 2 257
XM_011533391.2 XP_011531693.1 3 323
XM_011533392.2 XP_011531694.1 3 275
XM_011533394.2 XP_011531696.1 2 799
XM_011533396.2 XP_011531698.1 2 516
GenBank mRNA AK096749.1 - 2 140
AK295532.1 - 2 140
AK303160.1 - 2 154
BC020830.1 - 2 143 4771328
BM803652.1 - 2 87
CN430023.1 - 3 272
CV574708.1 - 4 393
DA952452.1 - 180
DC379144.1 - 2 165

View Full Product Details