Gene Transcripts

Gene Symbol
TMTC3
Entrez Gene ID
160418
Gene Name
transmembrane and tetratricopeptide repeat containing 3
Gene Aliases
SMILE
Chromosome Location
Chr. 12: 88142296 - 88199887 on Build GRCh38
UniGene ID
Hs.331268
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181783.3 NP_861448.2 5 799
XM_011537980.2 XP_011536282.1 4 574
GenBank mRNA AJ697717.1 - 5 669
AK074973.1 - 4 426
AK290436.1 - 5 799
AK302755.1 - 5 799
AK310375.1 - 5 799
AK310393.1 - 5 799
BC117175.1 - 4 620 40125726
BC117177.1 - 4 621 40125728
BX538169.1 - 5 772
CR749244.1 - 5 751
CR749309.1 - 5 785
CR933632.1 - 5 750

View Full Product Details