Gene Transcripts

Gene Symbol
ANKRD24
Entrez Gene ID
170961
Gene Name
ankyrin repeat domain 24
Gene Aliases
Chromosome Location
Chr. 19: 4182618 - 4224814 on Build GRCh38
UniGene ID
Hs.197872
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133475.1 NP_597732.1 12, 13 1235
XM_011527756.2 XP_011526058.1 13, 14 1145
XM_011527757.2 XP_011526059.1 12, 13 1393
XM_011527758.2 XP_011526060.1 12, 13 1058
XM_011527761.1 XP_011526063.1 14, 15 1215
XM_011527764.2 XP_011526066.1 11, 12 1306
XM_011527765.2 XP_011526067.1 11, 12 959
XM_017026438.1 XP_016881927.1 14, 15 1206
XM_017026439.1 XP_016881928.1 13, 14 1128
XM_017026440.1 XP_016881929.1 12, 13 1101
XM_017026441.1 XP_016881930.1 2, 3 243
GenBank mRNA AB075861.1 - 10, 11 932
BC028236.1 - 10, 11 877

View Full Product Details