Gene Transcripts

Gene Symbol
ABHD3
Entrez Gene ID
171586
Gene Name
abhydrolase domain containing 3
Gene Aliases
LABH3
Chromosome Location
Chr. 18: 21650897 - 21704805 on Build GRCh38
UniGene ID
Hs.397978
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138340.4 NP_612213.2 5 711
XM_011525847.2 XP_011524149.1 5 684
XM_017025571.1 XP_016881060.1 5 687
XM_017025572.1 XP_016881061.1 4 515
XM_017025573.1 XP_016881062.1 6 496
XM_017025574.1 XP_016881063.1 4 439
GenBank mRNA AF007152.1 - 5 670
AK024880.1 - 5 585
AK298728.1 - 5 610
BC021196.2 - 5 671 3942164

View Full Product Details