Gene Transcripts

Gene Symbol
KRT78
Entrez Gene ID
196374
Gene Name
keratin 78
Gene Aliases
CK-78, K5B, K78, Kb40
Chromosome Location
Chr. 12: 52837804 - 52848994 on Build GRCh38
UniGene ID
Hs.665267
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173352.3 NP_775487.2 1 185
XM_011538010.1 XP_011536312.1 1 185
GenBank mRNA AK096419.1 - 1 185
AK290901.1 - 1 185
AK298636.1 - 1 182
DA667429.1 - 1 186

View Full Product Details