Gene Transcripts

Gene Symbol
TMEM31
Entrez Gene ID
203562
Gene Name
transmembrane protein 31
Gene Aliases
Chromosome Location
Chr. X: 103710909 - 103714032 on Build GRCh38
UniGene ID
Hs.98843
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182541.2 NP_872347.2 3 391
GenBank mRNA AK130867.1 - 3 391
BC029575.1 - 3 392 5267530

View Full Product Details