Gene Transcripts

Gene Symbol
OR6T1
Entrez Gene ID
219874
Gene Name
olfactory receptor family 6 subfamily T member 1
Gene Aliases
OR11-277
Chromosome Location
Chr. 11: 123941474 - 123943844 on Build GRCh38
UniGene ID
Hs.553638
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005187.1 NP_001005187.1 1 308
XM_017017325.1 XP_016872814.1 2 223
GenBank mRNA BC136906.1 - 1 343

View Full Product Details