Gene Transcripts

Gene Symbol
MS4A15
Entrez Gene ID
219995
Gene Name
membrane spanning 4-domains A15
Gene Aliases
Chromosome Location
Chr. 11: 60756867 - 60776733 on Build GRCh38
UniGene ID
Hs.207465
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098835.1 NP_001092305.1 7 1814
NM_001278242.1 NP_001265171.1 6 1691
NM_152717.2 NP_689930.1 6 1527
NR_103481.1 8 2101
XM_006718459.3 XP_006718522.1 6 1534
XM_011544812.2 XP_011543114.1 7 1778
XM_011544814.1 XP_011543116.1 7 1643
GenBank mRNA AK091846.1 - 8 1964
AY584608.1 - 7 1673
AY584609.1 - 6 1550
BC031610.1 - 6 1391 5176452

View Full Product Details