Gene Transcripts

Gene Symbol
C6orf1
Entrez Gene ID
221491
Gene Name
chromosome 6 open reading frame 1
Gene Aliases
LBH
Chromosome Location
Chr. 6: 34246380 - 34249108 on Build GRCh38
UniGene ID
Hs.381300
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008703.2 NP_001008703.1 5 879
NM_001008704.2 NP_001008704.1 5 819
NM_001287396.1 NP_001274325.1 5 1071
NM_001287397.1 NP_001274326.1 4 1105
NM_001287398.1 NP_001274327.1 4 853
NM_178508.4 NP_848603.2 5 1131
GenBank mRNA AK290164.1 - 5 813
BC023627.2 - 5 751 4764944
BC047919.1 - 5 1012 6190218
BM474526.1 - 4 771
BU956518.1 - 4 618

View Full Product Details