Gene Transcripts
- Gene Symbol
- BRWD3
- Entrez Gene ID
- 254065
- Gene Name
- bromodomain and WD repeat domain containing 3
- Gene Aliases
- BRODL, MRX93
- Chromosome Location
- Chr. X: 80669488 - 80809736 on Build GRCh38
- UniGene ID
- Hs.147027
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153252.4 |
NP_694984.4
|
24, 25 |
3106 |
|
XM_005262113.3 |
XP_005262170.1
|
23, 24 |
2958 |
|
XM_017029384.1 |
XP_016884873.1
|
20, 21 |
2636 |
|
GenBank mRNA |
AK095887.1 |
- |
7, 8 |
979 |
|
AY497046.1 |
- |
24, 25 |
3106 |
|
AY497047.1 |
- |
21, 22 |
3038 |
|
AY497048.1 |
- |
24, 25 |
3129 |
|
AY497049.1 |
- |
23, 24 |
3099 |
|
AY497050.1 |
- |
24, 25 |
3079 |
|
AY497051.1 |
- |
21, 22 |
2666 |
|
AY497052.1 |
- |
26, 27 |
3265 |
|
AY497053.1 |
- |
22, 23 |
2854 |
|
AY497054.1 |
- |
21, 22 |
2755 |
|
AY497055.1 |
- |
24, 25 |
3040 |
|
AY497056.1 |
- |
21, 22 |
3011 |
|
AY497057.1 |
- |
20, 21 |
2910 |
|
AY497058.1 |
- |
20, 21 |
2749 |
|
AY497059.1 |
- |
22, 23 |
3120 |
|
AY497060.1 |
- |
18, 19 |
2687 |
|
View Full Product Details