Gene Transcripts

Gene Symbol
LSMEM1
Entrez Gene ID
286006
Gene Name
leucine rich single-pass membrane protein 1
Gene Aliases
C7orf53
Chromosome Location
Chr. 7: 112480219 - 112491062 on Build GRCh38
UniGene ID
Hs.396189
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134468.1 NP_001127940.1 4 465
NM_182597.2 NP_872403.1 4 671
XM_011516074.1 XP_011514376.1 4 1174
XM_011516075.2 XP_011514377.1 4 2861
XM_011516076.2 XP_011514378.1 4 2497
XM_017012028.1 XP_016867517.1 3 2379
GenBank mRNA AK096894.1 - 4 687
BC031976.1 - 6 738
BC126149.1 - 4 509 8991865
BC130568.1 - 4 509 40146599
DB456503.2 - 4 465

View Full Product Details