Gene Transcripts
- Gene Symbol
- STK11
- Entrez Gene ID
- 6794
- Gene Name
- serine/threonine kinase 11
- Gene Aliases
- LKB1, PJS, hLKB1
- Chromosome Location
- Chr. 19: 1205799 - 1228435 on Build GRCh38
- UniGene ID
- Hs.515005
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000455.4 |
NP_000446.1
|
6, 7 |
1984 |
|
XM_005259617.2 |
XP_005259674.1
|
6, 7 |
1494 |
|
XM_005259618.3 |
XP_005259675.1
|
6, 7 |
1494 |
|
XM_011528209.1 |
XP_011526511.1
|
7, 8 |
1625 |
|
GenBank mRNA |
AF035625.1 |
- |
6, 7 |
1207 |
|
AF217978.1 |
- |
7, 8 |
1830 |
|
AK314858.1 |
- |
6, 7 |
995 |
|
BC007981.2 |
- |
6, 7 |
1319 |
3689780 |
BC019334.1 |
- |
6, 7 |
1140 |
3946005 |
EF644172.1 |
- |
6, 7 |
953 |
|
KU178339.1 |
- |
6, 7 |
869 |
|
U63333.1 |
- |
6, 7 |
869 |
|
View Full Product Details