Gene Transcripts

Gene Symbol
ANKRD13D
Entrez Gene ID
338692
Gene Name
ankyrin repeat domain 13D
Gene Aliases
Chromosome Location
Chr. 11: 67289291 - 67302484 on Build GRCh38
UniGene ID
Hs.438673
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207354.2 NP_997237.2 10, 11 1244
NR_030767.1 11, 12 1327
GenBank mRNA AK027313.1 - 3, 4 1185
AK057681.1 - 3, 4 1858
AK096277.1 - 10, 11 1157
AK124721.1 - 11, 12 1324
AK126438.1 - 9, 10 1241
BC110419.1 - 10, 11 1217 5210878
BC121024.1 - 10, 11 1118 40117420
BC121025.2 - 10, 11 1118 40117423

View Full Product Details