Gene Transcripts
- Gene Symbol
- GOLGA6A
- Entrez Gene ID
- 342096
- Gene Name
- golgin A6 family member A
- Gene Aliases
- GLP, GOLGA6
- Chromosome Location
- Chr. 15: 74069857 - 74082550 on Build GRCh38
- UniGene ID
- Hs.546408
- Species
- Homo sapiens
- Gene Symbol
- GOLGA6B
- Entrez Gene ID
- 55889
- Gene Name
- golgin A6 family member B
- Gene Aliases
- GOLGA, GOLGA6D
- Chromosome Location
- Chr. 15: 72638291 - 72669604 on Build GRCh38
- UniGene ID
- Hs.546408
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018652.4 |
NP_061122.4
|
1 |
92 |
|
- Gene Symbol
- GOLGA6C
- Entrez Gene ID
- 653641
- Gene Name
- golgin A6 family member C
- Gene Aliases
- Chromosome Location
- Chr. 15: 75258404 - 75273455 on Build GRCh38
- UniGene ID
- Hs.540230
- Species
- Homo sapiens
- Gene Symbol
- GOLGA6D
- Entrez Gene ID
- 653643
- Gene Name
- golgin A6 family member D
- Gene Aliases
- Chromosome Location
- Chr. 15: 75282841 - 75295807 on Build GRCh38
- UniGene ID
- Hs.672474
- Species
- Homo sapiens
View Full Product Details