Gene Transcripts

Gene Symbol
CCDC18
Entrez Gene ID
343099
Gene Name
coiled-coil domain containing 18
Gene Aliases
NY-SAR-41
Chromosome Location
Chr. 1: 93179802 - 93279037 on Build GRCh38
UniGene ID
Hs.482873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306076.1 NP_001293005.1 15 2245
NM_206886.4 NP_996769.3 15 2248
XM_011541361.2 XP_011539663.2 15 2457
XM_011541372.2 XP_011539674.2 15 2457
XM_017001154.1 XP_016856643.1 15 2457
XM_017001155.1 XP_016856644.1 15 2454
XM_017001156.1 XP_016856645.1 15 2427
XM_017001157.1 XP_016856646.1 15 2409
XM_017001158.1 XP_016856647.1 15 2454
XM_017001159.1 XP_016856648.1 14 2325
XM_017001160.1 XP_016856649.1 15 2457
XM_017001161.1 XP_016856650.1 15 2457
XM_017001162.1 XP_016856651.1 15 2170
XM_017001163.1 XP_016856652.1 15 2724
XM_017001164.1 XP_016856653.1 15 2268
XM_017001165.1 XP_016856654.1 15 2457
XM_017001166.1 XP_016856655.1 13 1912
XM_017001167.1 XP_016856656.1 13 1847
XM_017001168.1 XP_016856657.1 11 1612
XM_017001169.1 XP_016856658.1 15 2457
XM_017001170.1 XP_016856659.1 2 1841
XM_017001171.1 XP_016856660.1 2 139
GenBank mRNA AK126045.1 - 9 1316
BX538286.1 - 7 1107

View Full Product Details