Gene Transcripts

Gene Symbol
KIAA1211L
Entrez Gene ID
343990
Gene Name
KIAA1211 like
Gene Aliases
C2orf55
Chromosome Location
Chr. 2: 98793846 - 98936221 on Build GRCh38
UniGene ID
Hs.469398
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207362.2 NP_997245.2 5, 6 832
XM_005263923.3 XP_005263980.1 5, 6 812
XM_005263924.3 XP_005263981.1 5, 6 812
XM_005263926.3 XP_005263983.1 5, 6 697
XM_011511093.1 XP_011509395.1 7, 8 971
XM_011511094.1 XP_011509396.1 6, 7 869
XM_011511095.2 XP_011509397.1 5, 6 639
XM_011511096.1 XP_011509398.1 2, 3 249
XM_017003971.1 XP_016859460.1 5, 6 815
XM_017003972.1 XP_016859461.1 6, 7 919
XM_017003973.1 XP_016859462.1 5, 6 825

View Full Product Details