Gene Transcripts

Gene Symbol
LCE2D
Entrez Gene ID
353141
Gene Name
late cornified envelope 2D
Gene Aliases
LEP12, SPRL1A
Chromosome Location
Chr. 1: 152663411 - 152664659 on Build GRCh38
UniGene ID
Hs.490225
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178430.3 NP_848517.1 2 453
GenBank mRNA BC130603.1 - 2 453
BC130605.1 - 2 453 40146636

View Full Product Details