Gene Transcripts

Gene Symbol
COA6
Entrez Gene ID
388753
Gene Name
cytochrome c oxidase assembly factor 6
Gene Aliases
C1orf31, CEMCOX4
Chromosome Location
Chr. 1: 234373437 - 234384049 on Build GRCh38
UniGene ID
Hs.23198
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012985.2 NP_001013003.1 1 81
NM_001301733.1 NP_001288662.1 1 63
GenBank mRNA BC116455.1 - 1 81

View Full Product Details