Gene Transcripts

Gene Symbol
MFSD2B
Entrez Gene ID
388931
Gene Name
major facilitator superfamily domain containing 2B
Gene Aliases
Chromosome Location
Chr. 2: 24009022 - 24026775 on Build GRCh38
UniGene ID
Hs.407482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080473.1 NP_001073942.1 7 729
XM_005264301.4 XP_005264358.1 7 1790
XM_017004063.1 XP_016859552.1 6 1883
XM_017004064.1 XP_016859553.1 6 1883
XM_017004065.1 XP_016859554.1 6 1883
XM_017004066.1 XP_016859555.1 6 1883
XM_017004067.1 XP_016859556.1 6 1883
XM_017004068.1 XP_016859557.1 6 1883
GenBank mRNA BX647146.1 - 2 151

View Full Product Details