Gene Transcripts

Gene Symbol
ARHGEF37
Entrez Gene ID
389337
Gene Name
Rho guanine nucleotide exchange factor 37
Gene Aliases
Chromosome Location
Chr. 5: 149552049 - 149634968 on Build GRCh38
UniGene ID
Hs.256206
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001669.2 NP_001001669.2 3, 4 372
XM_005268447.4 XP_005268504.1 3, 4 343
XM_005268448.1 XP_005268505.1 2, 3 246
XM_006714784.3 XP_006714847.1 4, 5 711
XM_011537642.2 XP_011535944.1 3, 4 394
XM_017009460.1 XP_016864949.1 3, 4 596
XM_017009461.1 XP_016864950.1 2, 3 246
GenBank mRNA AB199796.1 - 2, 3 319
AK300355.1 - 2, 3 233
AK300559.1 - 3, 4 471
AK316002.1 - 3, 4 442
BC041325.1 - 3, 4 372

View Full Product Details