Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163812.1 | NP_001157284.1 | 1 | 1415 | |
NM_021830.4 | NP_068602.2 | 1 | 1415 | ||
XM_017016437.1 | XP_016871926.1 | 1 | 914 | ||
GenBank mRNA | AF292004.1 | - | 1 | 925 | |
AF292005.1 | - | 1 | 925 | ||
AK025485.1 | - | 1 | 238 | ||
AK308098.1 | - | 1 | 1414 | ||
BC013349.2 | - | 1 | 348 | ||
BC033762.1 | - | 1 | 408 | 3854007 | |
BX640829.1 | - | 1 | 1397 |