Gene Transcripts

Gene Symbol
C19orf12
Entrez Gene ID
83636
Gene Name
chromosome 19 open reading frame 12
Gene Aliases
NBIA3, NBIA4, SPG43
Chromosome Location
Chr. 19: 29698886 - 29715789 on Build GRCh38
UniGene ID
Hs.529094
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031726.3 NP_001026896.2 3 418
NM_001256046.1 NP_001242975.1 3 689
NM_001256047.1 NP_001242976.1 3 495
NM_001282929.1 NP_001269858.1 2 248
NM_001282930.1 NP_001269859.1 2 519
NM_001282931.1 NP_001269860.1 4 810
NM_031448.4 NP_113636.2 3 689
GenBank mRNA AK057185.1 - 3 386
AL162066.1 - 2 193
BC004957.1 - 2 179 3628073
BC009946.2 - 2 179 4125477
BC017211.2 - 1 79
BC063518.1 - 3 368 4340755
BG720088.1 - 4 538
BI753584.1 - 3 421
DA348401.1 - 2 240
DA708831.1 - 3 411

View Full Product Details