Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004331.2 | NP_004322.1 | 4 | 522 | |
XM_005273617.3 | XP_005273674.1 | 4 | 426 | ||
GenBank mRNA | AB004788.1 | - | 4 | 443 | |
AF060922.1 | - | 4 | 476 | ||
AF067396.1 | - | 4 | 480 | ||
AF079221.1 | - | 4 | 480 | ||
AF452712.2 | - | 4 | 495 | ||
AF536326.1 | - | 4 | 458 | ||
AK290213.1 | - | 4 | 438 | ||
AK300286.1 | - | 4 | 447 | ||
AK307195.1 | - | 2 | 218 | ||
AK315870.1 | - | 4 | 399 | ||
AK316315.1 | - | 4 | 426 | ||
AL132665.1 | - | 4 | 496 | ||
BC001559.1 | - | 4 | 422 | 3459327 | |
BC009603.1 | - | 4 | 490 | 4134765 | |
BT019500.1 | - | 4 | 398 | ||
BT019501.1 | - | 4 | 398 | ||
CD558918.1 | - | 4 | 631 | ||
CR456701.1 | - | 4 | 398 |