Gene Transcripts

Gene Symbol
KIF7
Entrez Gene ID
374654
Gene Name
kinesin family member 7
Gene Aliases
ACLS, AGBK, HLS2, JBTS12, UNQ340
Chromosome Location
Chr. 15: 89627970 - 89663086 on Build GRCh38
UniGene ID
Hs.513134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198525.2 NP_940927.2 3 519
XM_011521531.2 XP_011519833.1 3 629

View Full Product Details