Gene Transcripts

Gene Symbol
SLC25A38
Entrez Gene ID
54977
Gene Name
solute carrier family 25 member 38
Gene Aliases
SIDBA2
Chromosome Location
Chr. 3: 39383324 - 39397328 on Build GRCh38
UniGene ID
Hs.369615
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017875.2 NP_060345.2 4, 5 856
XM_006713214.1 XP_006713277.1 5, 6 819
XM_011533869.2 XP_011532171.1 5, 6 1719
XM_017006710.1 XP_016862199.1 4, 5 855
GenBank mRNA AK000558.1 - 4, 5 832
AK225388.1 - 4, 5 808
BC013194.1 - 4, 5 517 3918744
CR457242.1 - 4, 5 455

View Full Product Details