Gene Transcripts
- Gene Symbol
- WDR19
- Entrez Gene ID
- 57728
- Gene Name
- WD repeat domain 19
- Gene Aliases
- ATD5, CED4, DYF-2, IFT144, NPHP13, ORF26, Oseg6, PWDMP, SRTD5
- Chromosome Location
- Chr. 4: 39182404 - 39285810 on Build GRCh38
- UniGene ID
- Hs.438482
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317924.1 |
NP_001304853.1
|
12, 13 |
1391 |
|
NM_025132.3 |
NP_079408.3
|
13, 14 |
1507 |
|
XM_011513724.1 |
XP_011512026.1
|
13, 14 |
1417 |
|
XM_011513725.2 |
XP_011512027.1
|
12, 13 |
1372 |
|
XM_011513726.2 |
XP_011512028.1
|
12, 13 |
1280 |
|
XM_011513727.1 |
XP_011512029.1
|
11, 12 |
1200 |
|
XM_017008501.1 |
XP_016863990.1
|
11, 12 |
1200 |
|
GenBank mRNA |
AB046858.1 |
- |
3, 4 |
351 |
|
AK294730.1 |
- |
12, 13 |
1266 |
|
AK304161.1 |
- |
13, 14 |
1380 |
|
AY029257.1 |
- |
13, 14 |
1507 |
|
View Full Product Details