Gene Transcripts

Gene Symbol
REN
Entrez Gene ID
5972
Gene Name
renin
Gene Aliases
HNFJ2
Chromosome Location
Chr. 1: 204154816 - 204166337 on Build GRCh38
UniGene ID
Hs.3210
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000537.3 NP_000528.1 6 809
GenBank mRNA BC033474.1 - 6 791
BC047752.1 - 6 794 5590211
CR536498.1 - 6 765
X00063.1 - 4 444

View Full Product Details