Gene Transcripts

Gene Symbol
ACBD7
Entrez Gene ID
414149
Gene Name
acyl-CoA binding domain containing 7
Gene Aliases
bA455B2.2
Chromosome Location
Chr. 10: 15075475 - 15088776 on Build GRCh38
UniGene ID
Hs.644598
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039844.2 NP_001034933.1 2, 3 173
GenBank mRNA AK095538.1 - 2, 3 173
BC029526.1 - 2, 3 173 5301753

View Full Product Details