Gene Transcripts
- Gene Symbol
- NBPF10
- Entrez Gene ID
- 100132406
- Gene Name
- neuroblastoma breakpoint family member 10
- Gene Aliases
- AB6, AG1, NBPF9
- Chromosome Location
- Chr. 1: 146066318 - 146144804 on Build GRCh38
- UniGene ID
- Hs.714127
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX538005.1 |
- |
11 |
1769 |
|
- Gene Symbol
- NBPF9
- Entrez Gene ID
- 400818
- Gene Name
- neuroblastoma breakpoint family member 9
- Gene Aliases
- AE01
- Chromosome Location
- Chr. 1: 149054033 - 149090861 on Build GRCh38
- UniGene ID
- Hs.445080
- Species
- Homo sapiens
View Full Product Details