Gene Transcripts

Gene Symbol
C8orf49
Entrez Gene ID
606553
Gene Name
chromosome 8 open reading frame 49
Gene Aliases
G4DM
Chromosome Location
Chr. 8: 11761256 - 11763223 on Build GRCh38
UniGene ID
Hs.545529
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_103552.1 1 678
GenBank mRNA AK055534.1 - 1 678
BC132945.1 - 1 547 40146967
BC136882.1 - 1 547 9020873

View Full Product Details