Gene Transcripts

Gene Symbol
AKAIN1
Entrez Gene ID
642597
Gene Name
A kinase (PRKA) anchor inhibitor 1
Gene Aliases
C18orf42
Chromosome Location
Chr. 18: 5143673 - 5199997 on Build GRCh38
UniGene ID
Hs.437160
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145194.1 NP_001138666.1 2 328
XM_011525728.2 XP_011524030.1 2 2655

View Full Product Details