Gene Transcripts

Gene Symbol
MBD5
Entrez Gene ID
55777
Gene Name
methyl-CpG binding domain protein 5
Gene Aliases
MRD1
Chromosome Location
Chr. 2: 148021011 - 148516923 on Build GRCh38
UniGene ID
Hs.458312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011511470.2 XP_011509772.1 2 279

View Full Product Details